MasterSense™ Smart Peristaltic Pumps
Designed for today's critical process applications and the biopharma industry of tomorrow
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
Monitor your process remotely via PC or mobile device.
It does not store programs or impact regulatory compliance.
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs.
More Pumping Options with Advanced Touchscreen Operation
Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping.
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.0001 to 68 ml/min *depending on cartridge type | Up to 35 psi | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
- Choose 1 or 2 channels
- Twist-and-open design, and customizable pump flow patterns ideal for laboratory use
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.004 to 365 ml/min | Up to 35 psi | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
Accurate and pulseless flows for pressurized and differential pressure applications and filling.
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 1.7 to 5850 ml/min | Up to 300 psi (20.7 bar) | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
Inert and accurate with touchscreen programmability for high pressure applications and dispensing sensitive or corrosive liquids.
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.025 to 75 ml/min | Up to 100 psi (6.9 bar) | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
直感的で革新的な
- 内蔵センサーが流体圧力を監視し、プロセス障害の発生前に警告を発します
- 標準プロトコルを保存・呼び出し、継続性を確保
- 直感的なデジタルタッチスクリーンで、最適化されたメニューナビゲーションとプログラム入力が可能 - 手袋着用時でも操作可能
- MasterSenseドライブは3段階のアクセス権限を提供し、異なるレベルのプログラミングを可能にします。すべてパスワード保護され、GMPベストプラクティスに準拠
接続されたスマートシステム
- 無線によるファームウェア更新で、メーカーへの返送なしにポンプシステムの最新ソフトウェアを確保
- MasterflexLive®クラウドアプリケーションが、すべてのMasterSense™ポンプシステムを安全に監視
- MasterflexLiveでは、稼働状態の変化やエラーメッセージを24時間365日通知します
簡単かつ正確
- 連続運転、容量ディスペンス、時間ディスペンスの運転モードを搭載
- 容量モードおよび時間モードにおけるバッチ制御と間隔/オフ時間
- 定性的なチューブキャリブレーションにより、チューブが精密なディスペンシングに最適化されていることを保証
- チューブが最適にキャリブレーションされた状態をグラフィカルインジケーターで表示
Compare MasterSense® Pumps
- Small footprint saves on benchtop space
- Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
- Add up to two L/S Pump heads, or shop Complete Pumps
- IP66 NEMA 4X stainless steel drives and complete systems
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 0.001 to 3400 ml/min | Up to 150 psi (10.3 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- All the features of the L/S MasterSense Drive, plus...
- 250 rpm max speed; dosing volumes from <0.5 ml to >500 ml
- Whisper-quiet operation
- Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life. Shop Double-Y tubing
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 0.0006 to 19 LPM | Up to 150 psi (10.3 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- Reliable pumps with the capacity and power for process applications
- Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000∶1 turndown ratio
- Add up to two I/P pump heads, or shop Complete Pumps
- IP66 NEMA 4X stainless steel drives and complete systems
HIGH FLOW |
PRESSURE |
CONNECTIVITY |
| 0.001 to 26 LPM | Up to 40 psi (2.7 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- IP66 washdown-rated 316 stainless steel housing
- Continuous-duty, reversible, variable-flow
- Choose PerfectPosition® Pump Tubing, B/T 87 or B/T 91
HIGHEST FLOW |
PRESSURE |
CONNECTIVITY |
| 0.65 to 42 LPM | Up to 35 psi | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |