ContactUsLinkComponent Orders Link
Mixing systems
Efficient Single-Use Mixing Systems
Advance your biomanufacturing with a comprehensive portfolio of single-use mixing systems. From shear-sensitive liquid-liquid or solid-liquid mixing to complex buffer or media applications, our standard, configurable and engineered solutions scale from 500 ml to 2,500 L with consistent, easy-to-use controls and 21 CFR 11-compliant software for seamless integration.
Mix with confidence and control
Image of an Avantor magnetic mixer
Tackle complex and shear-sensitive mixing with a magnetic drive and free-floating impeller in a sterile bag, scalable from 500 ml to 2,500 L with optional sensors and controls.
Image of an Avantor Quattro mixer
Enable gentle, aseptic mixing from 5 to 500 L with a Quattro mixer, recirculating fluid through a single-use path without moving parts to minimize shear, product damage and particle contamination.
Image of an Avantor top mixer
Optimize your bulk mixing with flexible open-top mixing systems, scalable up to 3,000 L. Compatible with a wide range of platforms, tank liners and accessories — ideal for efficient preparation of buffers and media.
Durable and easy to use, with an intuitive interface
Masterflex® single-use mixing systems combine usability, durability and process precision with unmatched expertise in fluid handling. As the only open-architecture fluid handling provider, we give you the freedom to integrate single-use components from an extensive, brand-agnostic portfolio or configure your hardware or software solutions to your exact process.
Our systems are built to streamline workflows, reduce contamination and scale efficiently. Intuitive controls and modular design help teams work more productively from lab to production.
Image highlighting and describing different features of Avantor mixing systems
Unparalleled quality in single-use mixing systems and consumables
Icon of a badge with a checkmark
Product testing & validation
Our single-use bags and assemblies undergo rigorous testing, including endotoxin, particulate and sterility validation, with documentation available on request to meet GMP standards.
Icon of a barcode
Lot traceability
We provide full component and manufacturing traceability, including production site, date and origin, to support compliance and simplify audits.
Icon of a cleanroom
ISO-certified cleanrooms
All Masterflex mixing assemblies are produced in ISO Class 7 cleanrooms within ISO 13485-certified, cGMP-compliant facilities to ensure product integrity.
Icon of a person
Our bioprocessing bags and single-use mixing systems are tailored to meet your specific application and regulatory requirements. A Masterflex single-use expert is ready to help you design a solution that accelerates your workflow and meets your production goals.
Featured resources
Two scientists in full protective gear testing and validating bioprocess bags
Review Masterflex’s validation guide to understand compliance, testing and manufacturing standards for single-use bioprocess bags.
Image showing some Avantor bioprocessing systems
Explore Masterflex’s full range of single-use systems, from mixing and filtration to buffer management, with standard, CTO and ETO options.
Image of several Avantor mixers
See how Masterflex’s magnetic mixers and assemblies helped scale viral vector and vaccine production while cutting CAPEX costs.
Learn how a retentate vessel with integrated single-use mixing modernized pilot-scale TFF workflows for scale-up.
Learn how a purpose-built retentate vessel with integrated single-use mixing modernized pilot-scale TFF workflows and supported reliable scale-up to commercial manufacturing.