ContactUsLinkComponent Orders Link
Custom bioprocess systems
Scalable Systems Tailored to Your Needs
Custom bioprocess systems meet pipeline, regulatory and capacity needs without sacrificing performance. Masterflex® is your one-stop shop delivering engineered-to-order (ETO) and configured-to-order (CTO) bioprocess solutions, including mixing, buffer preparation and TFF, all supported by single-use consumables and CFR 11-compliant automation.
Image of an Avantor mixing system
Scalable mixing with low shear control
Magnetic mixers from 50 mL to 2,500 L support gentle, high-performance blending for liquid-liquid or solids-liquid mixing with configured- and engineered-to-order flexibility.
Image of an Avantor tangential flow filtration system
TFF systems for purification
Engineer fully automated TFF systems for hollow fiber or cassette filters, designed to support crossflow filtration application from research to manufacturing with recirculation up to 5,000 L/h.
Image of an Avantor bioprocess system sitting on pump skids
Automated pumping and formulation
Design pump skids with integrated software and support for advanced control fluid transfer. Compatible with your choice of pump head, such as peristaltic, diaphragm or centrifugal.
Image of an Avantor inline dilution system
Dilution and conditioning on demand
Design inline dilution, conditioning or mixing systems for on-demand buffer or media preparation. Suitable for upstream and downstream processes with integrated controls.
A scientist adjusting tubing on an Avantor custom bioprocess system
Expertly engineered to your specifications
Masterflex custom bioprocess systems are designed using a structured process that begins with your requirements and ends with a fully integrated validated solution. Our global team manages everything from component selection and chassis design to documentation and integrated systems support. Whether you need a turnkey setup or a specialty system, we work closely with your team to deliver performance, compliance and scalability.
Trusted process, proven outcomes
Icon of a stylized globe with the letters
Certified quality & documentation
All systems are built to ISO 9001 standards, with full support for FAT/SAT, user manuals, optional IQ/OQ documentation and CFR 11-compliant software, GAMP 5 and ISA88.
Stylized icon showing a hexagon with a cloud upload system in the center
Flexible software integration
Open-architecture software connects upstream and downstream systems, supports SCADA, integrates with customer platforms and is CFR 11-compliant.
Icon of a person wearing a hardhat and a sheet of paper with some test tubes
Dedicated engineering support
A dedicated project manager oversees your system from design to post-delivery, working closely with your team to ensure performance and compliance.
Icon of a person with a checklist
Experience you can rely on
Masterflex has delivered over 850 systems worldwide, supporting small-scale builds to complex commercial installations. Backed by our cGMP manufacturing network, innovation centers and comprehensive supply capabilities.
custom-system-260.png
Start building your custom system
From concept to commissioning, our global team of engineers is ready to help design a solution tailored to your bioprocess. Whether you’re scaling up or optimizing an existing line, connect with us to get started.
Featured resources
A scientist working on a customized Avantor bioprocess system
Explore Masterflex’s full portfolio of single-use (SU) bioprocessing systems and mixing, filtration and fluid management solutions designed to scale with your process.
Image of an Avantor tangential flow filtration system
Learn how Masterflex’s partnership helped overcome TFF scale-up challenges with a custom SU system, improving yield, scalability and compliance.
Scientists in full protective gear in a buffer management facility
Find out how a hybrid buffer strategy model can help your team modernize buffer preparation to improve operational performance.