ContactUsLinkComponent Orders Link
cleanroom-1294339614-1920x760.jpg
Controlled Environment
Cleanrooms and other controlled environments used for scientific research or manufacturing pharmaceuticals, semiconductors and other electronic products require a host of specialized products. Protect your people and your products while maintaining a controlled environment – all from one source. Avantor® offers all the contamination control solutions you need to meet strict cleanroom and controlled environment compliance standards.
Controlled Environment and Cleanroom Solutions
New-Solutions-200x200.jpg
New Solutions
Find new products and suppliers in our controlled environment portfolio.
Solutions-by-Industry-200x200.jpg
Solutions by Industry
View environment control products from industry-leading manufacturers.
Featured-Brands-200x200.jpg
Controlled Environment Featured Brands
View environment control solutions from our featured suppliers.
Vendor-Services-200x200.jpg
Vendor Services
Let our vendors assist you with services to maintain a controlled and safe environment.
lightbulb-leaf-1298600057-955x690.jpg
Sustainable Solutions
Review our more ecofriendly safety and cleanroom supplies – either sustainably produced and packaged or recyclable
What is a Controlled Environment?
silicon-wafer-162267506-540x360.jpg
A controlled environment is one in which multiple environmental factors are monitored and controlled using controlled environment equipment. The controlled parameters typically include:
  • Particulates
  • Humidity
  • Temperature
  • Pressure
  • Segregation
What is a Cleanroom?
pharma-industry-1405590436-540x390.jpg
Cleanrooms are controlled environments that limit the number of pollutants, or contamination, to a specified number of particles (by size) per cubic foot or cubic meter. Cleanrooms are tested regularly to ensure standards are met and systems are in place to control the environment by limiting dust, chemical vapors, airborne microbes and aerosol particles.
Cleanrooms are used by manufacturing companies when a high level of sterility is necessary to avoid adversely affecting the product being manufactured. Cleanroom solutions vary based on the manufacturer’s needs and are used in a variety of industries, including:
  • Pharmaceutical
  • Biotech
  • Medical device
  • Life sciences
  • Aerospace
  • Semiconductor manufacturing
  • Optics
  • Military
  • Department of Energy
What Are the Standards for a Cleanroom?
cleanroom-microscope-1341292034-540x390.jpg
The standards for a cleanroom are based on the number of particles measured in one cubic foot or one cubic meter of air. Cleanrooms are classified using either the Federal Standard 209E of the USA or ISO 14644-1 standard.
Federal Standard 209E classifies cleanrooms with numbers like “class 1,000” or “class 10,000,” and classifications are based on the number of particles of size 0.5 μm or larger allowed in one cubic foot of air.
ISO 14644-1 standard classifies cleanrooms with smaller numbers like “3” and classifications are based on a decimal logarithm of the number of particles 0.1 μm or larger allowed in one cubic meter of air. 
Equipment and Supplies Necessary to Make Cleanrooms Meet Necessary Standards
cleanroom-pipetting-cell-culture-1166567136-540x390.jpg
The type of controlled environment lab equipment and supplies necessary to make sure a cleanroom meets necessary standards includes:
  • High-Efficiency Particulate Air (HEPA) filters
  • Ultra-Low Particulate Air (ULPA) filters
  • Airlocks
  • Air showers
  • Special gowning designed to trap contaminants
  • Environmental monitoring
Related content
controlled-env-workflow-solutions-540x390.jpg
The CES Ellipse showcases our contamination control solutions for the pharmaceutical, biotechnology, healthcare, microelectronics, aerospace, and automotive industries.
sustainable-production-supplies-540x390.jpg
Sustainable Solutions
Review our more ecofriendly safety and cleanroom supplies – either sustainability produced and packaged or recycled.
Are you ready to move your next innovation forward?
We can help. Contact us to speak with a member of our team.