MasterSense™ Smart Peristaltic Pumps
Designed for today's critical process applications and the biopharma industry of tomorrow
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
Monitor your process remotely via PC or mobile device.
It does not store programs or impact regulatory compliance.
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs.
More Pumping Options with Advanced Touchscreen Operation
Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping.
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.0001 to 68 ml/min *depending on cartridge type | Up to 35 psi | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
- Choose 1 or 2 channels
- Twist-and-open design, and customizable pump flow patterns ideal for laboratory use
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.004 to 365 ml/min | Up to 35 psi | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
Accurate and pulseless flows for pressurized and differential pressure applications and filling.
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 1.7 to 5850 ml/min | Up to 300 psi (20.7 bar) | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
Inert and accurate with touchscreen programmability for high pressure applications and dispensing sensitive or corrosive liquids.
LOW FLOW |
PRESSURE |
CONNECTIVITY |
| 0.025 to 75 ml/min | Up to 100 psi (6.9 bar) | EtherNet/IP, PROFINET, serial control, analog remote and MasterflexLive |
Intuitive and innovative
- Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
- Store and recall standard protocols, ensuring continuity
- Intuitive digital touchscreen with optimized menu navigation and program input — even with gloves on
- The MasterSense drives offer 3 tiers of access, allowing for varying levels of programming, all password protected, adhering to GMP best practices
Connected and smart
- Over-the-air firmware updates ensure your pump system has the latest software updates, without returning to the manufacturer
- MasterflexLive® cloud application securely monitors, all MasterSense pump systems
- With MasterflexLive, get notifications to alert you to changes in operating conditions and error messages 24/7
Easy and accurate
- Operational modes include continuous run, volume dispense and time dispense
- Batch control and interval/off time in volume and time modes
- Qualitative tube calibration ensures tubing is optimized for precise dispensing
- Graphical indicator lets you know when tubing is optimally calibrated
Compare MasterSense® Pumps
- Small footprint saves on benchtop space
- Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
- Add up to two L/S Pump heads, or shop Complete Pumps
- IP66 NEMA 4X stainless steel drives and complete systems
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 0.001 to 3400 ml/min | Up to 150 psi (10.3 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- All the features of the L/S MasterSense Drive, plus...
- 250 rpm max speed; dosing volumes from <0.5 ml to >500 ml
- Whisper-quiet operation
- Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life. Shop Double-Y tubing
MEDIUM FLOW |
PRESSURE |
CONNECTIVITY |
| 0.0006 to 19 LPM | Up to 150 psi (10.3 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- Reliable pumps with the capacity and power for process applications
- Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000∶1 turndown ratio
- Add up to two I/P pump heads, or shop Complete Pumps
- IP66 NEMA 4X stainless steel drives and complete systems
HIGH FLOW |
PRESSURE |
CONNECTIVITY |
| 0.001 to 26 LPM | Up to 40 psi (2.7 bar) | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |
- IP66 washdown-rated 316 stainless steel housing
- Continuous-duty, reversible, variable-flow
- Choose PerfectPosition® Pump Tubing, B/T 87 or B/T 91
HIGHEST FLOW |
PRESSURE |
CONNECTIVITY |
| 0.65 to 42 LPM | Up to 35 psi | EtherNet/IP, PROFINET, Profibus, serial control, analog remote and MasterflexLive |