ContactUsLinkComponent Orders Link
1271950_heroimage_1920x760.jpg
Reliable Bioprocess Containers for Secure Storage and Transport
Secure what matters. Protect the integrity of your process fluids and biological materials with Masterflex® bioprocess containers and 3D single-use bags engineered for secure storage and transport. Available in plastic and stainless steel, supporting volumes from 50 to 2,500 L.
Bioprocess containers and single-use bags for storage and transport
PlasticContainers-400x495 (1).jpg
Keep your single-use bioprocess bags secure during storage and transport with Masterflex's rigid plastic stackable containers. Designed for durability and flexibility, these containers can be collapsed for ease of storage, handling and shipment.
Image of an Avantor stainless steel container
Explore durable, stainless-steel containers, ideal for GMP environments and cleanroom transfer. Supporting working volumes of 50 to 1,500 L, these containers are easily configured with load cells can be double-jacketed for temperature control and have bag inflation options.
Image of a transport trolley carrying an Avantor plastic bin
Move heavy loads safely and efficiently with manual and motor-driven dollies and trolleys. Options include load cells and controlling software for advanced operational processing and quality control.
Image of an Avantor 3D single-use bioprocess bag
We offer standard 3D cubic bags in volumes from 50 to 1,500 L and custom bags up to 2,500 L. Our bioprocess bags have multiple port options constructed with a multi-layer film, providing product integrity and robust durability for storage and transport.
Engineered-to-order containers solutions
Engineered-to-order designs supporting volumes up to 2,500 L, with options for unique geometries and mobility requirements. In addition to standard configurable options, features like leakproof design and UV shielding are available.
A scientist in full protective gear pushing a trolley carrying an Avantor plastic bin
Protect quality from storage to delivery
Proper storage and transportation of process liquids and biological materials are essential for maintaining quality, safety and regulatory compliance. Bioprocess containers must perform reliably under pressure, temperature variation and movement, without compromising material integrity. Masterflex provides solutions designed to meet these demands and support consistent performance across your bioprocesses.
Proven performance backed by global expertise
Icon of a cleanroom
Validated for critical use
Our bioprocess containers are designed and tested to meet the demands of regulated environments, including ISTA and UN compatibility standards.
Icon of a person wearing a hard hat and a piece of paper with test tubes
Engineered for your processes
Standard and custom designs are available in stainless steel or polymer, with configurable features that match your process and compliance needs.
Icon of a globe with a package and arrow representing global support
Consistent supply worldwide
With global manufacturing and logistics support, Avantor® helps you maintain availability and continuity from pilot scale to commercial production.
Featured products
Image of several Avantor single-use mixers
Reduce contamination risk and improve efficiency with scalable, single-use mixing systems, configurable to your production requirements.
containers integrated offering_new.jpg
Design integrated, single-use or hybrid systems tailored to your processes. Masterflex’s custom assemblies and components support sterile fluid transfer, scale-up and cGMP compliance.
A scientist in full protective gear in a buffer management facility
Masterflex’s hydration platform simplifies solution preparation with critical reagent-grade buffers, concentrates and media in scalable formats.
Talk To An Expert
Whether you need standard bioprocess containers or customized designs, our team can help guide selection, validation and implementation. Connect with an expert to find the right fit for your process and compliance needs.
Featured resources
1658350_flyer_icon.jpg
Review product specifications, dimensions and compatibility information to help select the right storage and transport container for your bioprocess applications.
Icon of several reports
Discover our full range of single-use systems, from fluid handling and mixing to buffer management, all designed for scalable, compliant production.
Icon of a piece of paper with a magnifying glass
Review Avantor’s validation guide to understand compliance, testing and manufacturing standards for single-use bioprocess bags. 
Icon of a book
Access our complete library of brochures, datasheets and technical papers for bioprocess containers, assemblies and other single-use solutions.