J.T.Baker® Direct Dispense packaging system
Enhance the effectiveness of your products and streamline your biopharmaceutical manufacturing processes with the J.T.Baker® Direct Dispense packaging system. This novel packaging platform makes it easy to deliver powdered performance materials and excipients directly to your process, whether you are using traditional or single-use manufacturing equipment. Reduce time-consuming de-clumping and subdivision steps with products that are pre-weighed to your specifications and compatible with non-destructive Raman ID testing.
- Pre-weighed to your specifications
- Desiccants for reduced clumping
- Dispense tolerance of 1%
- Raman ID compatible with satellite sample
- Available in four sizes: 30 L, 65 L, 121 L, 150 L
Our experts discuss how the J.T.Baker® Direct Dispense packaging system is used in continuous biomanufacturing.
Avantor's J.T.Baker® Direct Dispense packaging system
In-time delivery of raw materials & excipients during continuous biomanufacturing
Learn how the J.T.Baker® Direct Dispense packaging system can make a difference in your operations.
Need to optimize your traditional or single-use packaging processes?
Learn how the J.T.Baker® Direct Dispense packaging system can make a difference in your operations.