France
ContactUsLinkComponent Lien pour commander
mflx-peri-pumps-1920x760.jpg
Masterflex® Peristaltic Pump Systems
Choose the right pump for you
Masterflex Pump Selector
Masterflex is an industry-leader in the design, engineering, and manufacturing of peristaltic tubing pumps. Coupled with precision-engineered tubing and hose pumps, diaphragm metering pumps, flowmeters, and mixers, Masterflex helps you develop a fluid handling ecosystem with the highest reproducibility and accuracy. Masterflex pumps are suited for many different types of fluid pathways, including :
  • Biopharma production
  • Academic / R&D - cell culture, filling/dispensing, filtration, general transfer
  • QA / QC laboratory sampling and analysis
  • Food & beverage process and dispensing
  • Water and wastewater treatment
  • Chemical process and manufacturing
  • Mineral testing and analysis
  • Environmental field and lab
  • Custom/OEM fluid handling solutions 
Masterflex offers you unparalleled versatility and standard-setting reliability – performance produced by more than 55 years of industry experience. Our selection of over 10,000 pump configurations alongside the expertise our team offers helps you find a pump system that meets the specific needs of your application.
Why Masterflex?
Masterflex® has set the worldwide standard in peristaltic pump systems. We have long been an industry leader in the designing, engineering and manufacturing of a wide variety of fluid handling products, including:
  • Peristaltic pump components and complete systems
  • Peristaltic pump tubing
  • Custom engineered pumps
Our expertise, plus our selection of over 10,000 pump configurations, means you get just the pump system you need. Over the course of more than 55 years of manufacturing peristaltic pumps we’ve long since proven that only Masterflex® offers such versatility and standard-setting reliability.
77920-40-42-77112-10-outlined-955.jpg
MasterSense™ smart pump systems
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Find your pump
Masterflex peristaltic pumps series advantages 
Masterflex peristaltic pumps are perfect for bioprocess, food & beverage process, laboratory/R&D, academic research, water treatment, chemical process, and other industrial and laboratory applications. They’re versatile, with wide flow ranges, flexible with several options for communication and control, and precise at fluid transfer and dispensing. And thanks to our proprietary design, they’re easy to clean and present little contamination risk to sensitive media and materials.
Top features:
  • Simple Process Validation - Fluid is completely contained in the tube and tubing is made from validated, biocompatible materials.
  • Cell Vitality/Molecular Integrity - Peristaltic pumps are low shear, reducing the physical impact on cells and large molecules, increasing overall cell vitality and molecular integrity for a more effective product/API.
  • Accurate & Linear Fluid Delivery - Positive displacement ensures high-accuracy volumetric flows across pressure ranges and viscosities in a linear relationship to rpm; the occlusion ensures no product leakage or backflow.
  • Masterflex® Exclusive: Fluid Path Ownership - Masterflex® offers control over almost every aspect of the fluid path such as pump, tubing, components, sensors, and the world class assembly services to bring it all together and deliver on your schedule. 
  • More reasons to choose Masterflex®- Self-priming and non-siphoning pumps ; 316SS housings, washdown and modular models; Variety of operator controls and connectivity; Safety and access security features
Tried, tested and trusted around the world:
Why Masterflex offers the peristaltic pumps of choice for countless applications
  1. Easy
    Quick installation. Low maintenance. Easy tubing changeover.
  2. Versatile
    From research to process, handles almost any fluid. Multichannel options for further fluid path configurability. 
  3. Accurate
    Dynamic flow ranges available, from 0.0001 mL/min to 42 L/min. Rigorous design standards for reproducible dispensing at any rate.
  4. Durable
    Trusted for over 55 years. Vast chemical compatibility.
  5. Customizable
    Configurable systems. Many tubing options. Easy scale up.
Avantor_Flowrate_GradientStroke-100.jpg
What flow rate do you need?
0.002 to 43 mL/min
Low flow pumps >
0.0006 to 3400 mL/min
Medium flow pumps >
0.001 to 42 LPM
High flow pumps >
Avantor_Chemical_Pumping_GradientStroke-100.jpg
What chemical are you pumping?
Match your chemicals to your materials
Chemical compatibility database >
Find your formula
Shop peristaltic tubing >
Avantor_Conditions_GradientStroke-100.jpg
What can influence my application?
Avantor_Connectivity_GradientStroke-100.jpg
What connectivity is necessary?
24/7 real-time remote monitoring
Get MasterflexLive >
Masterflex Digital Drive I/O
Masterflex Digital Drives Selection Guide >
touchscreen-icon-100.png
Intuitive and innovative
  • Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  • Store and recall standard protocols, ensuring continuity
  • Intuitive digital touchscreen with optimized menu navigation and program input — even with gloves on
  • The MasterSense drives offer 3 tiers of access, allowing for varying levels of programming, all password protected, adhering to GMP best practices
certified-icon-100.png
Easy and accurate
  • Operational modes include continuous run, volume dispense and time dispense
  • Batch control and interval/off time in volume and time modes
  • Qualitative tube calibration ensures tubing is optimized for precise dispensing
  • Graphical indicator lets you know when tubing is optimally calibrated
connectivity-icon-100.png
Connected and smart
  • Over-the-air firmware updates ensure your pump system has the latest software updates, without returning to the manufacturer
  • MasterflexLive® cloud application securely monitors, all MasterSense pump systems
  • With MasterflexLive, get notifications to alert you to changes in operating conditions and error messages 24/7
Masterflex offers the widest range of features, components, and flow rates to cover any fluid handling workflow
Peristaltic pump selection tools
service-mastersense-tool-100x100.jpg
Masterflex Selection Tool
Find the recommended bundled peristaltic pump system for you in less than 5 questions.
service-mastersense-configurator-100x100.jpg
Masterflex Configurator
Assemble your pump system by choosing features you need and resulting compatible components
Masterflex Peristaltic Pump Series — Shop by flow range
mflx-pump-series-LS.jpg
Precise flow control
Flow Range:
0.00005 to 3400 mL/min
Precision flow
mflx-pump-series-BT.jpg
Highest flow rates
Flow Range:
0.3 to 42 LPM
Highest flow pumps
IP_series_140x140_NEW.jpg
Powerful systems
Flow Range:
0.001 to 26 LPM
Industrial flow
mflx-pump-series-ISMATEC.jpg
Microflow, high accuracy
Flow Range:
0.0001 to 365 mL/min
Microflow pumps
undefined
Technical articles
undefined
Masterflex videos
Need more information or help selecting the perfect product for your application?
Our experts are ready to help. Contact us at 1-800-MASTERFLEX or [email protected].
Contact us